Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human MSP/MST1 (CHO-expressed) Protein, CF
SDP

Catalog No. 352MCH010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
352MCH010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 352MCH010 Supplier R&D Systems™ Supplier No. 352MCH010
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to activate MSP R/Ron in MDA-MB-453 or T47D human breast cancer cells. 80 ng/mL of the recombinant human MSP significantly induces phosphorylation of MSP R/Ron measured by the Human Phospho-MSP R/Ron DuoSet™ IC Kit (Catalog # DYC1947 ). Measured by its binding ability in a functional ELISA. When Recombinant Human MSPR/Ron Protein (Catalog # 1947-MS) is represent at 100 ng/mL (100 μg/mL), The concentration of Recombinant Human MSP/MST1 that produces 50% optimal binding response is found to be 8-40 ng/mL.

Specifications

Accession Number AAA59872
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 4485
Molecular Weight (g/mol) M.W. (Observed): 85 kDa, non-reducing conditions; M.W. (theoretical): 53 kDa (alpha-chain), 25 kDa (beta-chain)
Name MSP/MST1
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived human MSP/MST1 protein Gln19-Arg483 (alpha-chain) & Val484-Gly711 (beta-chain)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked heterodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol MST1
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein MSP/MST1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.