Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Nectin-4 Quantikine ELISA Kit
SDP

Catalog No. DNEC40 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DNEC40 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DNEC40 Supplier R&D Systems™ Supplier No. DNEC40

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Human Nectin-4 Microplate; Human Nectin-4 Conjugate; Human Nectin-4 Standard; Assay Diluent RD1-40; Calibrator Diluent RD5-26 Concentrate; Calibrator Diluent RD5-27; Wash Buffer Concentrate; Color Reagent A; Color Reagent B; Stop Solution; Plate Sealers

Human Nectin-4 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Nectin-4 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Nectin-4/PVRL4 in cell culture supernates (50 ul), cell lysates (25 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), urine (13 ul).

Specifications

Assay Range 39.10 to 2500 pg/mL
Assay Sensitivity 16.6 pg/mL
Conjugate HRP
Product Line R&D Systems
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human Nectin-4
For Use With (Application) ELISA
Gene ID (Entrez) 81607
Gene Symbol NECTIN4
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (25uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Urine (13uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym EDSS1, Ig superfamily receptor LNIR, LNIR, LNIRnectin 4, nectin-4, Nectin4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4, PRR4DKFZp686K05193, PVRL4
Target Nectin-4/PVRL4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.