Learn More
R&D Systems™ Human Pepsinogen A Recombinant Protein
Description
Source: Mouse myeloma cell line, NS0-derived, Met1-Ala388, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 Âμg per lane.
Molecular Weight: 41 kDa
Formulation: Supplied as a 0.2μm filtered solution in Tris and NaCl.
Entrez Gene ID: 643847
Regulatory Status: RUO
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 643847 |
| Molecular Weight (g/mol) | 41 kDa |
| Name | Pepsinogen A |
| Quantity | 10 μg |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.