Learn More
Description
Includes
Mouse anti-CD9, Mouse anti-E-Cadherin, Goat anti-Nanog, Goat anti-Oct-3/4, Mouse anti-PODXL (GCTM antigen), Mouse anti-SOX2, Mouse anti-SSEA-1, Mouse anti-SSEA-4
Eight markers for increased confidence in pluripotency status: anti-CD9, anti-E-Cadherin, anti-Nanog, anti-Oct-3/4, anti-PODXL (GCTM antigen), anti-SOX2, anti-SSEA-1, and anti-SSEA-4
Verifies the pluripotency of your starting stem cell population
Cost-e
Specifications
Specifications
| Cross Reactivity | Human |
| Quantity | 1 Kit |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Form | Lyophilized |
| Product Line | R&D Systems |
| Product Type | Neural Progenitor Markers |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
