Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Proprotein Convertase 1/PCSK1 Protein, CF

Catalog No. 2810SE010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2810SE010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2810SE010 Supplier R&D Systems™ Supplier No. 2810SE010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave the fluorogenic peptide substrate pERTKR-AMC (Catalog # ES013 ). The specific activity is >30 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P29120
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 5122
Molecular Weight (g/mol) M.W. (Observed): 63-80 kDa, reducing conditions; M.W. (theoretical): 57 kDa
Name Proprotein Convertase 1/PCSK1
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived human Proprotein Convertase 1/PCSK1 protein Ser111-Arg617, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Structural Form Mature form
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol PCSK1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Proprotein Convertase 1/PCSK1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.