Learn More
R&D Systems™ Human S5a/Angiocidin Biotin Recombinant Protein, E. coli-derived
Description
Specifications
Specifications
| Formulation | Supplied as a solution in HEPES, NaCl, DTT and Glycerol. |
| Gene ID (Entrez) | 5710 |
| Name | S5a/Angiocidin Biotin |
| Quantity | 250μg |
| Source | E. coli-derived human S5a/Angiocidin protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Biotin |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.