Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Serpin C1/Antithrombin-III Quantikine ELISA Kit
SDP

Catalog No. DSPC10 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DSPC10 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DSPC10 Supplier R&D Systems™ Supplier No. DSPC10
Add to Cart
Add to Cart
This item is not returnable. View return policy

a 3.5 hour solid-phase ELISA designed to measure human Serpin C1 in cell culture supernates, serum, plasma, urine, and human milk.

The Human Serpin C1/Antithrombin-III Quantikine ELISA Kit quantifies human Serpin C1/Antithrombin-III in cell culture supernates (25 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), citrate plasma (10 ul), urine (25 ul), human milk (10 ul).

Specifications

Assay Range 0.31 - 20ng/mL
Assay Sensitivity 0.097ng/mL
Detectable Analytes Serpin C1/Antithrombin-III
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity Cross-species reactivity observed with 1 or more species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well Strip Plate
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Citrate, Heparin), Serum, Urine
Specificity Natural and Recombinant free and complexed Human Serpin C1. This assay cross-reacts 1.4% with Recombinant Mouse Serpin C1.
For Use With (Application) ELISA
Gene ID (Entrez) 462
Host Species Human
Measuring Range 0.313-20ng/mL
Quantity 1 Kit
Sample Volume Cell Culture Supernates (25uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Citrate Plasma (10uL),Urine (25uL),Human Milk (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Antithrombin-III, AT3antithrombin-III, ATIII, ATIIIantithrombin III, MGC22579, serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1, serine-cysteine proteinase inhibitor clade C member 1, Serpin C1, serpin peptidase inhibitor, clade C (antithrombin), member 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.