Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human SUMO E1 (SAE1/UBA2) Protein, CF

Catalog No. E315025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E315025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E315025 Supplier R&D Systems™ Supplier No. E315025
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Recombinant Human SUMO E1 (SAE1/UBA2) is a member of the SUMO-activating (E1) enzyme family that is required for the first step of the enzymatic cascade that subsequently utilizes a SUMO-conjugating (E2) enzyme to conjugate SUMO to substrate proteins. A SUMO ligase (E3) is sometimes utilized for SUMO conjugation, but is not always required. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human SUMO E1 (SAE1/UBA2) concentration of 50-500 nM.

Specifications

Accession Number Q9UBEO (SAE1) & Q9UBT2 (UBA2)
For Use With (Application) Enzyme Activity
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 10055
Molecular Weight (g/mol) M.W. (theoretical): 38 kDa (SAE1) & 71 kDa (UBA2)
Name SUMO Activating Enzyme E1 (SAE1/UBA2)
Quantity 25 μg
Source E. coli-derived human SUMO Activating Enzyme E1 (SAE1/UBA2) protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Symbol SAE1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Protein SUMO Activating Enzyme E1 (SAE1)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.