Learn More
R&D Systems™ Human SUMO1 Rhodamine Recombinant Protein
Description
Specifications
Specifications
| Formulation | Xmg/mL (XμM) in 50mM HEPES pH 7.5, 100mM NaCl. |
| Gene ID (Entrez) | 7341 |
| Molecular Weight (g/mol) | Predicted MW: 12kDa |
| Name | SUMO1 Rhodamine |
| Quantity | 50μg |
| Source | E. coli-derived human SUMO1 protein |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Rhodamine |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by PAGE. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.