Learn More
R&D Systems™ Human SUMO1 Vinyl Sulfone C-Terminal Derivative Recombinant Protein
Description
Specifications
Specifications
| Accession Number | P63165 |
| Formulation | X mg/ml (X μM) in 50 mM MES pH 6.5, 100 mM NaCl |
| Gene ID (Entrez) | 7341 |
| Molecular Weight (g/mol) | Predicted MW: 11kDa |
| Name | SUMO1 Vinyl Sulfone C-Terminal Derivative |
| Quantity | 50μg |
| Source | E. coli-derived human SUMO1 protein Contains a C-terminal Vinyl Sulfone |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Vinyl Sulfone |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.