Learn More
R&D Systems™ Human SUMO2 Agarose Recombinant Protein
Description
Specifications
Specifications
| Formulation | 250μl of agarose supplied as 500μl total volume in 50mM HEPES pH 8, 250mM NaCl, 0.09% sodium azide |
| Gene ID (Entrez) | 6613 |
| Name | SUMO2 Agarose |
| Quantity | 500 μL |
| Source | E. coli-derived human SUMO2 protein |
| Storage Requirements | Do not freeze. 3 months from date of receipt, 2 to 8° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. |
| Conjugate | Agarose |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.