Learn More
R&D Systems™ Human SUMO3 Agarose Recombinant Protein
Description
Specifications
Specifications
| Formulation | Formulation cannot be determined without the BulkLotPrefix |
| Gene ID (Entrez) | 6612 |
| Name | SUMO3 Agarose |
| Quantity | 500 μL |
| Source | E. coli-derived human SUMO3 protein |
| Storage Requirements | Do not freeze. 3 months from date of receipt, 2 to 8° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. |
| Conjugate | Agarose |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.