Learn More
R&D Systems™ Human SUMO3 Biotin Recombinant Protein
Description
Specifications
Specifications
| Formulation | No Formulation Info in Bulk Inventory |
| Gene ID (Entrez) | 6612 |
| Name | SUMO3 Biotin |
| Quantity | 50 μg |
| Source | E. coli-derived human SUMO3 protein |
| Storage Requirements | Store the unopened product at -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Conjugate | Biotin |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by PAGE. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.