Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Syndecan-1 Protein, CF

Catalog No. 2780SD050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2780SD050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2780SD050 Supplier R&D Systems™ Supplier No. 2780SD050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by the ability of the immobilized protein to enhance the adhesion of Saos-2 human osteosarcoma cells to human Fibronectin. Measured by the ability of the immobilized protein to enhance the adhesion of Saos-2 human osteosarcoma cells to human Fibronectin (0.5 μg/mL, Catalog # 4305-FNB ). The ED50 for this effect is 2.5-10 μg/mL.

Specifications

Accession Number NP_002988
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 6382
Molecular Weight (g/mol) M.W. (Observed): 60-90 kDa, reducing conditions; M.W. (theoretical): 24.9 kDa
Name Syndecan-1/CD138
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human Syndecan-1/CD138 protein Gln18-Glu251, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol SDC1
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Syndecan-1/CD138
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.