Learn More
R&D Systems™ Human Tetra-Ubiquitin/Ub4 Non-hydrolyzable (K63), Agarose Recombinant Protein
Description
Specifications
Specifications
| Accession Number | P0CG47 |
| Formulation | No Formulation Info in Bulk Inventory |
| Gene ID (Entrez) | 7314 |
| Name | Tetra-Ubiquitin/Ub4 Non-hydrolyzable (K63), Agarose |
| Quantity | 100 μL |
| Source | E. coli-derived human Tetra-Ubiquitin protein Each Ubiquitin contains a Pro substitution at position 73 |
| Storage Requirements | Do not freeze. 3 months from date of receipt, 2 to 8° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. |
| Conjugate | Agarose |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.