Learn More
R&D Systems™ Human Tetra-Ubiquitin/Ub4 Non-hydrolyzable (K63) Recombinant Protein
Description
Specifications
Specifications
| Accession Number | P0CG47 |
| Formulation | Lyophilized from deionized water. Reconstitute at 2mg/mL in aqueous buffer. |
| Gene ID (Entrez) | 7314 |
| Name | Tetra-Ubiquitin/Ub4 Non-hydrolyzable (K63) |
| Quantity | 25μg |
| Source | E. coli-derived human Tetra-Ubiquitin protein Each Ubiquitin contains a Pro substitution at position 73 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.