Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Tetra-Ubiquitin/Ub4 Non-hydrolyzable (Linear) Recombinant Protein

Catalog No. UCN710100
Change view
Click to view available options
100ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
UCN710100 100ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. UCN710100 Supplier R&D Systems™ Supplier No. UCN710100
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Linear Tetra-Ub Non-hydrolyzable is derived from E. coli. The Recombinant Human Linear Tetra-Ub Non-hydrolyzable has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number P0CG47
Formulation Lyophilized from deionized water. Reconstitute at 2mg/mL in aqueous buffer.
Gene ID (Entrez) 7314
Name Tetra-Ubiquitin/Ub4 Non-hydrolyzable (Linear)
Quantity 100μg
Source E. coli-derived human Tetra-Ubiquitin protein Each Ubiquitin contains a Pro substitution at position 73
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.