Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human THOP1 Protein, CF

Catalog No. 3439ZN010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3439ZN010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 3439ZN010 Supplier R&D Systems™ Supplier No. 3439ZN010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave a fluorogenic peptide substrate, (7-methoxycoumarin-4-yl)acetyl-Pro-Leu-Gly-Pro-D-Lys(2,4-dinitrophenyl)-OH or Mca-PLGPK(Dnp)-OH. The specific activity is >250 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number NP_003240
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in MES, NaCl and Glycerol.
Gene ID (Entrez) 7064
Molecular Weight (g/mol) M.W. (Observed): 80 kDa, reducing conditions; M.W. (theoretical): 80 kDa
Name Thimet Oligopeptidase/THOP1
Quantity 10 μg
Source E. coli-derived human Thimet Oligopeptidase/THOP1 protein Lys2-Cys689, with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol THOP1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Thimet Oligopeptidase/THOP1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.