Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Thrombospondin-1 Protein, CF
SDP

Catalog No. 3074TH050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3074TH050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 3074TH050 Supplier R&D Systems™ Supplier No. 3074TH050

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by the ability of the immobilized protein to support the adhesion of SVEC4-10 mouse vascular endothelial cells. When 2 x 104 cells/well are added to DTT treated THBS-1 coated plates (10 μg/mL, 100 μL/well), > 50% will adhere after 1 hour at 37 °C.

Specifications

Accession Number P07996-1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in TBS, NaCl, and Trehalose.
Gene ID (Entrez) 7057
Molecular Weight (g/mol) M.W. (Observed): 135 - 165 kDa, reducing conditions; M.W. (theoretical): 129 kDa
Name Thrombospondin-1
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human Thrombospondin-1 protein Asn19-Pro1170, with Thr523Ala substitution and a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol THBS1
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Thrombospondin-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.