Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Total Serpin E1/PAI-1 Quantikine ELISA Kit
SDP

Catalog No. DTSE100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DTSE100 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DTSE100 Supplier R&D Systems™ Supplier No. DTSE100
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Cell Culture Supernates (50μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Citrate Plasma (10μL)

Human Total Serpin E1/PAI-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Total Serpin E1/PAI-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Serpin E1/PAI-1 in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), citrate plasma (10 ul).

Specifications

Assay Range 0.31 - 20ng/mL
Assay Sensitivity 0.046ng/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human Total Serpin E1 (active, latent, uPA, tPA, and Vitronectin complexed Serpin E1).
Target Species Human
For Use With (Application) ELISA
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Citrate Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Endothelial plasminogen activator inhibitor, Nexin, PAI1, PAI-1, PAI1PAI-1, PAISerpin E1, PLANH1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.