Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human TREM-1 Quantikine ELISA Kit
SDP

Catalog No. DTRM10C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DTRM10C 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DTRM10C Supplier R&D Systems™ Supplier No. DTRM10C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human TREM-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human TREM-1 Quantikine ELISA Kit quantifies human TREM-1 in cell culture supernates (50 ul), cell lysates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), saliva (50 ul).

Specifications

Assay Range 62.50 - 4000pg/mL
Assay Sensitivity 15.2pg/mL
Detectable Analytes TREM-1
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well Strip Plate
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Saliva, Serum
Specificity Natural and Recombinant Human TREM-1
For Use With (Application) ELISA
Gene ID (Entrez) 54210
Host Species Human
Measuring Range 62.5-4,000pg/mL
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Saliva (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CD354, CD354 antigen, TREM1, TREM-1 Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
Target TREM1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.