Learn More
R&D Systems™ Human Tri-Ubiquitin/Ub3 Non-hydrolyzable (K48), Agarose Recombinant Protein
Description
Specifications
Specifications
| Accession Number | P0CG47 |
| Formulation | 100 μl of agarose supplied in a 200 μl total volume of 20% Ethanol. |
| Gene ID (Entrez) | 7314 |
| Name | Tri-Ubiquitin/Ub3 Non-hydrolyzable (K48), Agarose |
| Quantity | 100 μL |
| Source | E. coli-derived human Tri-Ubiquitin protein Each Ubiquitin contains a Pro substitution at position 73 |
| Storage Requirements | Do not freeze. 3 months from date of receipt, 2 to 8° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. |
| Conjugate | Agarose |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.