Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human UbcH8/UBE2L6 Protein, CF

Catalog No. E2644100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E2644100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E2644100 Supplier R&D Systems™ Supplier No. E2644100
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Recombinant Human UbcH8/UBE2L6 is a member of the Ub AND ISG15-conjugating (E2) enzyme family that receives Ub AND ISG15 from a Ub AND ISG15-activating (E1) enzyme and subsequently interacts with a Ub AND ISG15 ligase (E3) to conjugate Ub AND ISG15 to substrate proteins. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human UbcH8/UBE2L6 concentration of 0.1-1 μM.

Specifications

Accession Number O14933
For Use With (Application) Enzyme Activity
Formulation X mg/mL (X μM) in 50 mM HEPES pH 7.5, 100 mM NaCl, 10% Glycerol (v/v), 1 mM TCEP
Gene ID (Entrez) 9246
Molecular Weight (g/mol) M.W. (theoretical): 18 kDa
Name UbcH8
Quantity 100 μg
Source E. coli-derived human UbcH8 protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Symbol UBE2L6
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Protein UbcH8/Ube2L6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.