Learn More
R&D Systems™ Human Ubiquitin Aldehyde C-Terminal Derivative Recombinant Protein
Description
Specifications
Specifications
| Formulation | Xmg/mL (XμM) as a solution in 50mM HCl |
| Gene ID (Entrez) | 7314 |
| Molecular Weight (g/mol) | 8.6 kDa |
| Name | Ubiquitin Aldehyde C-Terminal Derivative |
| Quantity | 50μg |
| Source | E. coli-derived human Ubiquitin protein Contains a C-terminal Aldehyde |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Aldehyde |
| Recombinant | Recombinant |
| Purity or Quality Grade | 96%, by HPLC. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.