Learn More
R&D Systems™ Human Ubiquitin Mutant I44A Recombinant Protein
Description
Specifications
Specifications
| Formulation | No Formulation Info in Bulk Inventory |
| Gene ID (Entrez) | 7314 |
| Name | Ubiquitin Mutant I44A |
| Quantity | 1mg |
| Source | E. coli-derived human Ubiquitin protein |
| Storage Requirements | Store the unopened product at 2 - 8degrees C. Do not use past expiration date. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.