Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Ubiquitin Mutant with K27 only Recombinant Protein

Catalog No. UMK27001M
Change view
Click to view available options
1mg; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
UMK27001M 1mg; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. UMK27001M Supplier R&D Systems™ Supplier No. UMK27001M
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Ubiquitin Mutant with K27 only Protein is derived from E. coli. The Recombinant Human Ubiquitin Mutant with K27 only Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number P0CG47
Formulation Lyophilized from a solution in sterile, deionized water
Gene ID (Entrez) 7314
Molecular Weight (g/mol) Predicted MW: 8.7kDa
Name Ubiquitin Mutant with K27 only
Quantity 1mg
Source E. coli-derived human Ubiquitin protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.