Learn More
R&D Systems™ Human Ubiquitin Mutant K48R Recombinant Protein
Description
Specifications
Specifications
| Accession Number | P0CG47 |
| Formulation | Lyophilized from a solution in deionized water. |
| Gene ID (Entrez) | 7314 |
| Molecular Weight (g/mol) | Predicted MW: 8.6kDa |
| Name | Ubiquitin Mutant K48R |
| Quantity | 1mg |
| Source | E. coli-derived human Ubiquitin protein Contains a Lys-to-Arg substitution at position 48. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.