Learn More
R&D Systems™ Human Ubiquitin Mutant with K63 only, N-Terminal Biotin Recombinant Protein
Description
Specifications
Specifications
| Formulation | No Formulation Info in Bulk Inventory |
| Gene ID (Entrez) | 7314 |
| Name | Ubiquitin Mutant with K63 only, N-Terminal Biotin |
| Quantity | 50μg |
| Source | E. coli-derived human Ubiquitin protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Biotin |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by PAGE. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.