Learn More
R&D Systems™ Human Ubiquitin Vinyl Methyl Ester C-Terminal Derivative Recombinant Protein
Description
Specifications
Specifications
| Formulation | 2.15mg/mL (250μM) in 50mM MES pH 6.0 |
| Gene ID (Entrez) | 7314 |
| Name | Ubiquitin Vinyl Methyl Ester C-Terminal Derivative |
| Quantity | 50μg |
| Source | E. coli-derived human Ubiquitin protein Contains underivatized and C-terminal Vinyl Methyl Ester derivatized protein,quantity is by derivatized content |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Vinyl Methyl Ester |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by HPLC. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.