Learn More
R&D Systems™ Human UFM1 Agarose Recombinant Protein
Description
Specifications
Specifications
| Formulation | 0.5 ml Ufm1 agarose is supplied in a 1 ml total volume of 50 mM Hepes pH 7.5, 250 mM NaCl. |
| Gene ID (Entrez) | 51569 |
| Name | UFM1 Agarose |
| Quantity | 500μL |
| Source | E. coli-derived human UFM1 protein |
| Storage Requirements | Do not freeze. 3 months from date of receipt, 2 to 8° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. |
| Conjugate | Agarose |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.