Learn More
R&D Systems™ Human UFM1 Rhodamine Recombinant Protein
Description
Specifications
Specifications
| Formulation | X mg/mL (X μM) in 50 mM Hepes pH 7.5, 100 mM NaCl. Actual concentration varies with lot number |
| Gene ID (Entrez) | 51569 |
| Name | UFM1 Rhodamine |
| Quantity | 50μg |
| Source | E. coli-derived human UFM1 protein |
| Storage Requirements | Store the unopened product at -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Conjugate | Rhodamine |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by PAGE. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.