Learn More
R&D Systems™ Human URM1 Isoform 1 Recombinant Protein
Description
Specifications
Specifications
| Formulation | X mg/ml (X μM) in 50 mM HEPES pH 7.5, 100 mM NaCl, 1 mM DTT |
| Gene ID (Entrez) | 81605 |
| Name | URM1 Isoform 1 |
| Quantity | 250μg |
| Source | E. coli-derived human URM1 protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.