Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human vWF-A2 DuoSet ELISA
SDP

Catalog No. DY276405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY276405 1 Kit (for 5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY276405 Supplier R&D Systems™ Supplier No. DY276405
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)

Human vWF-A2 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human vWF-A2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human vWF-A2.

Specifications

Assay Range 46.90 to 3000 pg/mL
Detectable Analytes vWF-A2
Assay Solid Phase Sandwich ELISA
Conjugate Biotin
Cross Reactivity <50%cross-species reactivity observed with species tested
Immunoassay Kit Format 96-well Strip Plate
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant
Sufficient For To run ELISAs on at least 5 x 96-well plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 7450
Gene Symbol VWF
Host Species Human
Quantity 1 Kit (for 5 Plates)
Sample Volume 100 μL
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym F8VWF, VWD, VWF von Willebrand factor, vWFA2
Target vWF-A2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.