Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human XCL1/Lymphotactin Quantikine ELISA Kit
SDP

Catalog No. DXCL10
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DXCL10 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DXCL10 Supplier R&D Systems™ Supplier No. DXCL10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Human Lymphotactin microplate, 96-well; Human Lymphotactin conjugate; Human Lymphotactin standard; Buffers and diluents, Color reagents A and B; Stop solution; Plate sealers

A 4.5 hour solid-phase sandwich ELISA designed to measure human Lymphotactin in cell culture supernates, serum, and plasma

The Human XCL1/Lymphotactin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human XCL1/Lymphotactin in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).

Specifications

Quantity 1 Kit
Species Human
Conjugate HRP
Assay Sensitivity 0.023 ng/mL
Assay Range 0.16 to 10 ng/mL
For Use With (Application) ELISA
Sample Type Cell Culture Supernates (100uL), Serum (100uL), EDTA Plasma (100uL), Heparin Plasma (100uL)
Synonym ATAC, ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1 alpha, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin)
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.