Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hyaluronan (Low Molecular Weight), R&D Systems™
SDP

Catalog No. GLR001
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GLR001 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. GLR001 Supplier R&D Systems Supplier No. GLR001

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material Hyaluronan
Color White
Physical Form White odorless powder
Synonym Hyaluronic acid (HA), Sodium hyaluronate
Quantity 100 mg
Formula Weight Predicted MW: 15 - 40kDa Observed MW: 20.0kDa
Odor Odorless
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.