Learn More
R&D Systems™ MagCellect Human CD3+ T Cell Isolation Kit
SDP
Catalog No. MAGH101
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
MAGH101 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. MAGH101 Supplier R&D Systems™ Supplier No. MAGH101
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Designed to recover 45 to 70% of target cells via negative selection principle with a 90-98% purity
A mononuclear cell suspension is first incubated with the MagCellect Antibody Cocktail which targets the unwanted cells. MagCellect Streptavidin Ferrofluid is next added to the reaction which allows the streptavidin coated nanoparticles to interact with biotinylated antibody tagged cells. The tube containing the cell suspension is then placed within a magnetic field. Magnetically tagged cells will migrate toward the magnet (unwanted cell fraction), leaving the untagged cells or desired cell population in suspension to be harvested by aspiration while the tube remains in the magnetic field.
Designed for the isolation of cells in a liquid phase
Based on the use of Ferrofluids or magnetic nanoparticles that have no magnetic memory (superparamagnetic) and behave as colloids
Generates particles with higher ligand binding capacity per mass compared to many other larger diameter (>150nm) magnetic particles
The enriched cell preparation is then available for a variety of applications including tissue culture, immune status monitoring and flow cytometry
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit