Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Malachite Green Phosphate Detection Kit
SDP

Catalog No. DY996 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY996 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY996 Supplier R&D Systems™ Supplier No. DY996
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

contains sufficient materials to run Sandwich ELISAs on at least 1 Kit

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits

Specifications

Product Line R&D Systems
Product Type Malachite Green Phosphate Detection Kit
Sufficient For Fifteen 96-well plates
For Use With (Application) Phosphate Detection
Kit Contents Malachite Green Reagent A (Ammonium molybdate in 3M sulfuric acid); Malachite Green Reagent B (Malachite green oxalate and polyvinyl alcohol); Phosphate Standard
Quantity 1 Kit
Sample Volume 50 μL
Storage Requirements Store at room temperature
Total Assay Time 4 hours 40 minutes (after plate preparation)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.