Learn More
Description
It is also an excellent substrate for Trypsin, HGF activator and factor Xa.
Specifications
Specifications
| Form | Liquid |
| For Use With (Application) | Bioactivity, Binding Activity |
| Product Line | R&D Systems |
| Quantity | 1 mg |
| Format | Supplied as a stock solution in dimethyl sulfoxide (DMSO) at a concentration of 10.0mg/mL or 4.3 mM. |
| Molecular Weight (g/mol) | Predicted MW: 1674 Da |
| Product Type | Purity: >95%, by HPLC, Source: Synthetic |
| Storage Requirements | Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
