Learn More
R&D Systems™ Mca-RPPGFSAFK(Dnp)-OH Fluorogenic Peptide Substrate

Description
- Contains a highly fluorescent 7-methoxycoumarin group that is efficiently quenched by resonance energy transfer to the 2, 4-dinitrophenyl group
- It can be used to measure the activities of peptidases that are capable of cleaving an amide bond between the fluorescent group and the quencher group, causing an increase in fluorescence
- Purity >95% by HPLC
- The cleavage site by ECE-1 is the peptide bond between Ala and Phe
Specifications
Specifications
| Content And Storage | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. |
| Form | Liquid |
| For Use With (Application) | Bioactivity, Binding Activity |
| Quantity | 1 mg |
| Format | Stock solution at 4mM or 6.26mg/mL (0.16 mL) dimethyl sulfoxide (DMSO). |
| Molecular Weight (g/mol) | Predicted MW: 1388 Da |
| Product Type | Source: Synthetic |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.