Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MK6-83
GREENER_CHOICESDP

Catalog No. 554750
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
554750 50 mg
554710 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 554750 Supplier Tocris Bioscience™ Supplier No. 5547/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TRPML channel activator

MK6-83 is a TRPML channel activator (EC50 = 110 nM). Also activates F408Δ and F465L TRPML mutants (EC50 values are 0.1 and 1.23 μM respectively). Restores endolysosomal trafficking and zinc homeostasis in lysosomes of mucolipidosis type IV (MLIV) mutant fibroblasts.

Specifications

Chemical Name or Material MK6-83
CAS 1062271-24-2
Synonym 5-Methyl-N-[2-(1-piperidinyl)phenyl]-2-thiophenesulfonamide
Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Molecular Formula C16H20N2O2S2
Formula Weight 336.47
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.