Learn More
R&D Systems™ Mouse ADAM9 Recombinant Protein
Description
Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala206-Asp697, Ser, and a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 55 kDa
Formulation: Supplied as a 0.2μm filtered solution in Tris, NaCl, ZnCl2, Brij-35 and Glycerol.
Entrez Gene ID: 11502
Regulatory Status: RUO
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl, ZnCl2, Brij-35 and Glycerol. |
| Gene ID (Entrez) | 11502 |
| Molecular Weight (g/mol) | 55 kDa |
| Name | ADAM9 |
| Quantity | 20 μg |
| Structural Form | Mature form. Recombinant Mouse ADAM9 is prone to proteolytic cleavage at C-terminus. The poly-His tag may not be present in the preparation. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.