Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse C-Reactive Protein/CRP Quantikine ELISA Kit
GSA_VASDP

Catalog No. MCRP00 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MCRP00 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MCRP00 Supplier R&D Systems™ Supplier No. MCRP00
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Mouse CRP Microplate; Mouse CRP Standard; Mouse CRP Control; Mouse CRP Conjugate; Assay Diluent RD1W; Calibrator Diluent RD5P Concentrate; Wash Buffer Concentrate; Color Reagent A; Color Reagent B; Stop Solution; Plate Sealers

For the quantitative determination of mouse C-Reactive Protein (CRP) concentrations in cell culture supernates, tissue lysates, serum, and plasma

The Mouse C-Reactive Protein/CRP Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse C-Reactive Protein/CRP in cell culture supernates (50 ul), tissue lysates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Assay Range 0.16 - 10ng/mL
Assay Sensitivity 0.015ng/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Lysates
Specificity Natural and Recombinant Mouse CRP
For Use With (Application) ELISA
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Tissue Lysates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.