Learn More
R&D Systems™ Mouse CD3+ T Cell Enrichment Column, Small
Description
CD3+ is a universal marker for T cells.
Total T cell recovery from these columns ranged between 45% and 65%
Purity (CD3+ cells) of recovered cells ranged between 81% and 88%
Based upon gravitational flow
Simple procedure
No specialized equipment required
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Includes | Murine T Cell Selection Columns, 10X Column Buffer Concentrate |
| Quantity | 10/Pk. |
| Type | Cell Enrichment Column |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.