Learn More
R&D Systems™ Mouse CD4+ T Cell Enrichment Column, Mini
Description
CD4 is an approximately 55kDa type I transmembrane glycoprotein that is expressed predominantly on thymocytes and a subset of mature T cells. In humans, CD4 is also expressed to a lesser extent on monocytes and macrophage related cells.
Recovery of CD3+/4+ cells from total CD3+/4+ cells loaded ranged between 20% and 32%
Purity of recovered cells ranged between 86% and 92% with no detectable CD3+/8+ cells
Leukocyte suspensions are incubated with a mixture of monoclonal antibodies and then loaded onto T Cell Subset Columns
B cells, nonselected T cells and monocytes bind to glass beads coated with anti-immunoglobulin via both F(ab) and Fc interactions
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Quantity | 4/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.