Learn More
R&D Systems™ Mouse CD8+ T Cell Enrichment Column, Small
Description
CD8 is a heterodimeric glycoprotein consisting of an alpha and beta chain. It is expressed on cytolytic T cells and functions in conjunction with the T cell receptor in the recognition of MHC/peptide complexes.
Recovery of CD3+/8+ cells from all available CD3+/8+ cells loaded ranged between 34% and 52%
Purity of recovered cells ranged between 80% and 88% with no detectable CD3+/4+ cells in the recovered population
Monocyte cell suspensions are incubated with a mixture of monoclonal antibodies and then loaded onto T Cell Subset Columns
B cells, nonselected T cells and monocytes bind to glass beads coated with anti-immunoglobulin via both F(ab) and Fc interactions
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Quantity | 4/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.