Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse CTLA-4 Fc Chimera Protein
GSA_VA

Catalog No. 434CT200
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
434CT200 Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
434CT200CF Lyophilized from a 0.2 μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 434CT200 Supplier R&D Systems™ Supplier No. 434CT200
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit IL-2 secretion by stimulated Jurkat human acute T cell leukemia cells. Linsley, P.S. et al. (1991) J. Exp. Med. 174:561. The ED50 for this effect is 0.1-0.4 μg/mL when stimulated with 1 μg/mL Recombinant Human B7-1/CD80 Fc Chimera (Catalog # 140-B1 ) in the presence of PHA.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Accession Number Q6GTR6
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 1493
Molecular Weight (g/mol) M.W. (Observed): 55 kDa, reducing conditions; M.W. (theoretical): 41 kDa (monomer)
Name CTLA-4 Fc Chimera
Quantity 200 μg
Source Mouse myeloma cell line, NS0-derived mouse CTLA-4 protein Mouse CTLA-4 (Ala37-Phe162) (N-terminus) Accession #Q6GTR6 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Ctla4
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein CTLA-4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.