Learn More
R&D Systems™ Mouse Erythrocyte Lysing Kit

Description
- Gently lyses erythrocytes
- Stabilizes leukocytes for analysis by flow cytometry
- Maintains natural light scattering and fluorescent staining properties of leukocytes
- Maintains viability of leukocytes for subsequent tissue culture manipulation
- Gently lyses erythrocytes
- Stabilizes leukocytes for analysis by flow cytometry
- Maintains natural light scattering and fluorescent staining properties of leukocytes
- Maintains viability of leukocytes for subsequent tissue culture manipulation
Specifications
Specifications
| Includes | M-Lyse Buffer Concentrate (10X); Wash Buffer Concentrate (10X); Fixative Concentrate (10X) |
| Product Type | Cell Lysis Kit |
| Content And Storage | Store at 20°C to 25°C |
| Product Line | R&D Systems |
| For Use With (Application) | Removal of erythrocytes (red blood cells) from whole blood |
| Sufficient For | 12.5 x 109 splenocytes |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.