Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Granzyme B ELISpot Development Module, 5 Plate
SDP

Catalog No. SEL1865 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SEL1865 1 Kit (for 5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SEL1865 Supplier R&D Systems™ Supplier No. SEL1865

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sufficient for at least five 96-well plates

  • ELISpot development module provides optimized capture and detection antibody pairings for the quantitative sandwich ELISpot detection of mouse IFN-γ.
  • It is an economical alternative to complete kits that allows for assay customization and reduced development time.
  • Assay Type: Quantitative sandwich ELISpot development module.
  • Species Reactivity: Mouse.
  • Assay Capacity: Five 96-well microplates.
  • Included Components: Lyophilized anti-mouse IFN-γ capture antibody and lyophilized biotinylated anti-mouse IFN-γ detection antibody.
  • Recommended Plates: Nitrocellulose-bottom plates, PVDF-bottom plates, or flat-bottom polystyrene ELISA plates.Research Use: For research use only, not for use in diagnostic procedures.

Specifications

Antigen Granzyme B
Cross Reactivity Mouse
Quantity 1 Kit (for 5 Plates)
Synonym C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTLA-1, CTSGL1, Fragmentin-2, granzyme B, Granzyme-2, GranzymeB, GRB, GrzB, GZMB, HLP, SECT
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Gene ID (Entrez) 3002
Gene Symbol Gzmb
Product Line R&D Systems
Host Species Mouse
Product Type ELISpot Development Module
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.