Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse His6-UCH-L1/PGP9.5 Protein, CF

Catalog No. E335025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E335025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E335025 Supplier R&D Systems™ Supplier No. E335025
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Recombinant Mouse His6-UCH-L1 is a Ubiquitin-specific deconjugating enzyme. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Mouse His6-UCH-L1 concentration of 10-50 nM. A 15 minute pre-incubation with 10 mM DTT is recommended to achieve maximum activity.

Specifications

Accession Number Q9R0P9
For Use With (Application) Enzyme Activity
Formulation X mg/mL (X μM) in 50 mM HEPES, pH 8.0, 2 mM DTT
Gene ID (Entrez) 7345
Molecular Weight (g/mol) M.W. (theoretical): 27 kDa
Name His6 UCH-L1
Quantity 25 μg
Source E. coli-derived mouse UCH-L1/PGP9.5 protein Contains a 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Symbol Uchl1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Protein UCH-L1/PGP9.5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.