Learn More
R&D Systems™ Recombinant Mouse IFN-beta Protein
Catalog No. 124011
Click to view available options
:
100000U, Carrier-Free
This item is not returnable. View return policy
Catalog No. 124011
Supplier: R&D Systems™ 124011
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Specifications
Storage Requirements Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Gene Alias Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.